Product Details

SNP ID
rs74106202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:95578919 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGTGGCACATCCATGTTAAGGG[A/G]CTGAGGCGTCCCTGGCACGGAATGC
Phenotype
MIM: 608671
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLDN10 PubMed Links

Gene Details

Gene
CLDN10
Gene Name
claudin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160100.1 6774 UTR 3 NP_001153572.1
NM_006984.4 6774 UTR 3 NP_008915.1
NM_182848.3 6774 UTR 3 NP_878268.1
XM_011521134.2 6774 Intron XP_011519436.1
XM_017020843.1 6774 Intron XP_016876332.1
XM_017020844.1 6774 Intron XP_016876333.1
Gene
DZIP1
Gene Name
DAZ interacting zinc finger protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014934.4 6774 UTR 3 NP_055749.1
NM_198968.3 6774 UTR 3 NP_945319.1
XM_005254032.1 6774 Intron XP_005254089.1
XM_006719916.1 6774 Intron XP_006719979.1
XM_006719917.2 6774 Intron XP_006719980.1
XM_006719918.1 6774 Intron XP_006719981.1
XM_006719920.1 6774 Intron XP_006719983.1
XM_017020444.1 6774 Intron XP_016875933.1
XM_017020445.1 6774 Intron XP_016875934.1
XM_017020446.1 6774 Intron XP_016875935.1
XM_017020447.1 6774 Intron XP_016875936.1

View Full Product Details