Product Details

SNP ID
rs73406680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52310198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTTTTTGTGCTTCTGCACCTGA[C/T]AAAAGAAGCATCAGATATGTGGAAA
Phenotype
MIM: 160777
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYO5A PubMed Links

Gene Details

Gene
MYO5A
Gene Name
myosin VA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000259.3 9112 UTR 3 NP_000250.3
NM_001142495.1 9112 UTR 3 NP_001135967.1
XM_005254397.3 9112 UTR 3 XP_005254454.1
XM_011521606.2 9112 UTR 3 XP_011519908.2
XM_011521607.2 9112 UTR 3 XP_011519909.2
XM_011521608.2 9112 UTR 3 XP_011519910.2
XM_011521609.2 9112 UTR 3 XP_011519911.2
XM_011521610.2 9112 UTR 3 XP_011519912.2
XM_011521611.2 9112 UTR 3 XP_011519913.2
XM_011521612.2 9112 UTR 3 XP_011519914.2
XM_017022227.1 9112 UTR 3 XP_016877716.1

View Full Product Details