Product Details

SNP ID
rs72758928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64365485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCAACCGTTTAAACCCTCAAGA[A/C]CAAATTATCTTTACATTTTTTTAAA
Phenotype
MIM: 606274 MIM: 610696
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CSNK1G1 PubMed Links

Gene Details

Gene
CSNK1G1
Gene Name
casein kinase 1 gamma 1
There are no transcripts associated with this gene.

Gene
KIAA0101
Gene Name
KIAA0101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029989.2 857 UTR 3 NP_001025160.1
NM_014736.5 857 UTR 3 NP_055551.1

View Full Product Details