Product Details

SNP ID
rs67877911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:69837265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAATCAAGATAAGTATTATTTCAA[A/T]GCAGTATATGTAAAAATTGAAAGTA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00593 PubMed Links

Gene Details

Gene
LINC00593
Gene Name
long intergenic non-protein coding RNA 593
There are no transcripts associated with this gene.

View Full Product Details