Product Details

SNP ID
rs71399777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74175147 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGCCGCCGGAGCCCTGGCCTCT[C/T]TGAGCCATCTCAAGAGCCTGGACCT
Phenotype
MIM: 602059 MIM: 610745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ISLR PubMed Links

Gene Details

Gene
ISLR
Gene Name
immunoglobulin superfamily containing leucine rich repeat
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005545.3 571 Silent Mutation CTG,TTG L97L NP_005536.1
NM_201526.1 571 Silent Mutation CTG,TTG L97L NP_958934.1
Gene
STRA6
Gene Name
stimulated by retinoic acid 6
There are no transcripts associated with this gene.

View Full Product Details