Product Details

SNP ID
rs74035978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:96784190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTTCTCAGAAGCCATGACATGT[A/C]CCTGCGGCTGGAGGCCTTTCAAGGG
Phenotype
MIM: 613948
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPATA8 PubMed Links

Gene Details

Gene
SPATA8
Gene Name
spermatogenesis associated 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304804.1 221 Missense Mutation ACC,CCC T4P NP_001291733.1
NM_001304805.1 221 Missense Mutation ACC,CCC T4P NP_001291734.1
NM_001304806.1 221 Missense Mutation ACC,CCC T4P NP_001291735.1
NM_173499.4 221 Missense Mutation ACC,CCC T43P NP_775770.1
XM_011521265.2 221 Missense Mutation ACC,CCC T4P XP_011519567.1
Gene
SPATA8-AS1
Gene Name
SPATA8 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details