Product Details
- SNP ID
-
rs73551016
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:25220222 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCTCAACTGAATGTCCTATGTCC[G/T]TTGGAGGATGGGTAAGCTCATTGGT
- Phenotype
-
MIM: 603750
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
AQP8
PubMed Links
Gene Details
- Gene
- AQP8
- Gene Name
- aquaporin 8
View Full Product Details