Product Details

SNP ID
rs72813578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88858574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATAGTTCAGAGCTGGTGTGCGC[C/T]GGGTGGAGTCTTGTCCTTTCAGTCG
Phenotype
MIM: 612222 MIM: 610970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GALNS PubMed Links

Gene Details

Gene
GALNS
Gene Name
galactosamine (N-acetyl)-6-sulfatase
There are no transcripts associated with this gene.

Gene
PABPN1L
Gene Name
poly(A) binding protein nuclear 1 like (cytoplasmic)
There are no transcripts associated with this gene.

Gene
TRAPPC2L
Gene Name
trafficking protein particle complex 2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318524.1 Intron NP_001305453.1
NM_001318525.1 Intron NP_001305454.1
NM_001318526.1 Intron NP_001305455.1
NM_001318527.1 Intron NP_001305456.1
NM_001318528.1 Intron NP_001305457.1
NM_001318529.1 Intron NP_001305458.1
NM_001318530.1 Intron NP_001305459.1
NM_001318532.1 Intron NP_001305461.1
NM_016209.4 Intron NP_057293.1
XM_017023270.1 Intron XP_016878759.1

View Full Product Details