Product Details

SNP ID
rs73343804
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:73335213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTTTGGTTTGTCTTGGCAGTGT[A/G]GGTGTGGAGCTGACGTCAACTCTGG
Phenotype
MIM: 607217
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SDK2 PubMed Links

Gene Details

Gene
SDK2
Gene Name
sidekick cell adhesion molecule 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144952.1 9893 UTR 3 NP_001138424.1
XM_011524914.2 9893 UTR 3 XP_011523216.1
XM_011524915.2 9893 UTR 3 XP_011523217.1
XM_011524916.2 9893 Intron XP_011523218.1

View Full Product Details