Product Details

SNP ID
rs73999366
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82060026 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCTTGGACAGGACCTCCGTGC[C/T]ACCCTCCTCTTCCTCCAGGGCCCGC
Phenotype
MIM: 601934 MIM: 602578
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DUS1L PubMed Links
Additional Information
For this assay, SNP(s) [rs202150137] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DUS1L
Gene Name
dihydrouridine synthase 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022156.4 1252 Missense Mutation AGC,GGC S364G NP_071439.3
XM_005256393.2 1252 Missense Mutation AGC,GGC S364G XP_005256450.1
XM_005256394.2 1252 Missense Mutation AGC,GGC S364G XP_005256451.1
XM_006722288.2 1252 Missense Mutation AGC,GGC S347G XP_006722351.1
XM_006722289.2 1252 Missense Mutation AGC,GGC S347G XP_006722352.1
Gene
GPS1
Gene Name
G protein pathway suppressor 1
There are no transcripts associated with this gene.

Gene
RFNG
Gene Name
RFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
There are no transcripts associated with this gene.

View Full Product Details