Product Details

SNP ID
rs74006125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81899261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAGGTCCTCTCCATGGAGAAAG[C/G]ATTTCTAGGTGTTTGGGCTGGTGCC
Phenotype
MIM: 604171 MIM: 614534 MIM: 607996 MIM: 602679
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ALYREF PubMed Links

Gene Details

Gene
ALYREF
Gene Name
Aly/REF export factor
There are no transcripts associated with this gene.

Gene
ANAPC11
Gene Name
anaphase promoting complex subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002244.2 291 Silent Mutation AGC,AGG S43R NP_001002244.1
NM_001002245.2 291 Intron NP_001002245.1
NM_001002246.2 291 Intron NP_001002246.1
NM_001002247.2 291 Intron NP_001002247.1
NM_001002248.2 291 Intron NP_001002248.1
NM_001002249.2 291 Intron NP_001002249.1
NM_001289414.1 291 Intron NP_001276343.1
NM_001289415.1 291 Intron NP_001276344.1
NM_001289416.1 291 Intron NP_001276345.1
NM_001289417.1 291 Intron NP_001276346.1
NM_001289418.1 291 Intron NP_001276347.1
NM_001289419.1 291 Intron NP_001276348.1
NM_001289420.1 291 Silent Mutation AGC,AGG S43R NP_001276349.1
NM_016476.11 291 Intron NP_057560.8
Gene
NPB
Gene Name
neuropeptide B
There are no transcripts associated with this gene.

Gene
PCYT2
Gene Name
phosphate cytidylyltransferase 2, ethanolamine
There are no transcripts associated with this gene.

View Full Product Details