Product Details

SNP ID
rs73274366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15930939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGGAACTTGACAAGTAAGCCTA[A/C]ATTCACTGGGAAGGATGAAGCTAAG
Phenotype
MIM: 600446
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADORA2B PubMed Links

Gene Details

Gene
ADORA2B
Gene Name
adenosine A2b receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000676.2 Intron NP_000667.1
XM_011523659.2 Intron XP_011521961.1
XM_011523661.2 Intron XP_011521963.1
XM_017024197.1 Intron XP_016879686.1

View Full Product Details