Product Details

SNP ID
rs72835771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:6641214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCGGGGGGAAAAGCTGGTGCCC[C/G]GACTGCGTGCAGGGTGAGGCCGGGA
Phenotype
MIM: 616967
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0753 PubMed Links

Gene Details

Gene
KIAA0753
Gene Name
KIAA0753
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014804.2 313 Intron NP_055619.2
XM_006721612.2 313 Intron XP_006721675.1
XM_011524090.2 313 Intron XP_011522392.1
XM_011524091.2 313 Intron XP_011522393.1
XM_011524095.2 313 Intron XP_011522397.1
XM_011524096.1 313 Intron XP_011522398.1
XM_011524098.2 313 Intron XP_011522400.1
XM_011524099.2 313 Intron XP_011522401.1
XM_011524100.2 313 Intron XP_011522402.1
XM_017025455.1 313 Intron XP_016880944.1
XM_017025456.1 313 Intron XP_016880945.1
XM_017025457.1 313 Intron XP_016880946.1
Gene
MED31
Gene Name
mediator complex subunit 31
There are no transcripts associated with this gene.

Gene
TXNDC17
Gene Name
thioredoxin domain containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032731.3 313 Silent Mutation CCC,CCG P44P NP_116120.1

View Full Product Details