Product Details

SNP ID
rs72846981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8732609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATAATAAGTGGTCACATGCCCCA[A/G]AGGAGTCCTGTGGTCAAGTCAGTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC42 PubMed Links

Gene Details

Gene
CCDC42
Gene Name
coiled-coil domain containing 42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001158261.1 Intron NP_001151733.1
NM_144681.2 Intron NP_653282.2

View Full Product Details