Product Details

SNP ID
rs72829744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:56120990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCGATTTTTCCCATTTCCCTGACA[C/T]TTTAACCTATATCCTAGTCTTCCTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKFN1 PubMed Links

Gene Details

Gene
ANKFN1
Gene Name
ankyrin repeat and fibronectin type III domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153228.2 Intron NP_694960.2
XM_006721728.3 Intron XP_006721791.1
XM_011524428.2 Intron XP_011522730.1
XM_011524429.2 Intron XP_011522731.1
XM_011524430.2 Intron XP_011522732.1
XM_011524431.2 Intron XP_011522733.1
XM_011524433.2 Intron XP_011522735.1
XM_011524434.2 Intron XP_011522736.1
XM_017024263.1 Intron XP_016879752.1
XM_017024264.1 Intron XP_016879753.1
XM_017024265.1 Intron XP_016879754.1
XM_017024266.1 Intron XP_016879755.1
XM_017024267.1 Intron XP_016879756.1
XM_017024268.1 Intron XP_016879757.1
XM_017024269.1 Intron XP_016879758.1
XM_017024270.1 Intron XP_016879759.1

View Full Product Details