Product Details

SNP ID
rs73331103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59566904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCAGTATTTGCTTTATAGTCCC[A/G]CTTCTCTTGGCACACCCCCTTCCCC
Phenotype
MIM: 607570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHX40 PubMed Links

Gene Details

Gene
DHX40
Gene Name
DEAH-box helicase 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166301.1 Intron NP_001159773.1
NM_024612.4 Intron NP_078888.4
XM_006722083.2 Intron XP_006722146.1
XM_011525253.2 Intron XP_011523555.1
XM_011525254.2 Intron XP_011523556.1
XM_017025080.1 Intron XP_016880569.1
XM_017025081.1 Intron XP_016880570.1
XM_017025082.1 Intron XP_016880571.1
XM_017025083.1 Intron XP_016880572.1

View Full Product Details