Product Details

SNP ID
rs73340967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:67075938 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCCTTATCCAAAGCTTCCATCA[A/T]CAACATATTTATATAAGGTGATGGG
Phenotype
MIM: 606699
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HELZ PubMed Links

Gene Details

Gene
HELZ
Gene Name
helicase with zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014877.3 8330 UTR 3 NP_055692.2
XM_005257888.4 8330 UTR 3 XP_005257945.1
XM_005257889.4 8330 UTR 3 XP_005257946.1
XM_006722214.3 8330 UTR 3 XP_006722277.1
XM_006722215.3 8330 UTR 3 XP_006722278.1
XM_006722216.3 8330 UTR 3 XP_006722279.1
XM_011525544.2 8330 UTR 3 XP_011523846.1
XM_017025477.1 8330 UTR 3 XP_016880966.1
XM_017025478.1 8330 UTR 3 XP_016880967.1

View Full Product Details