Product Details

SNP ID
rs73335250
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:49711589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAGAGACACACAAGACACATA[C/T]GTACACACAGAGAGAAACAGACAGC
Phenotype
MIM: 610790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM117A PubMed Links

Gene Details

Gene
FAM117A
Gene Name
family with sequence similarity 117 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030802.3 Intron NP_110429.1
XM_017025182.1 Intron XP_016880671.1
Gene
SLC35B1
Gene Name
solute carrier family 35 member B1
There are no transcripts associated with this gene.

View Full Product Details