Product Details

SNP ID
rs73290783
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:36067394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGATGTTTGGCACAACTTATAAC[A/C]ATGAAAAATTGGCCAGGTGCAGTGG
Phenotype
MIM: 603757
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCL18 PubMed Links

Gene Details

Gene
CCL18
Gene Name
C-C motif chemokine ligand 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002988.3 Intron NP_002979.1

View Full Product Details