Product Details

SNP ID
rs73401859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:26024143 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTTAAATGTACTGAATAGGCAA[A/G]TCTATAGAGATAGAAAGTAAACTAC
Phenotype
MIM: 600192
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SS18 PubMed Links

Gene Details

Gene
SS18
Gene Name
SS18, nBAF chromatin remodeling complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007559.2 Intron NP_001007560.1
NM_001308201.1 Intron NP_001295130.1
NM_005637.3 Intron NP_005628.2
XM_006722527.2 Intron XP_006722590.1
XM_011526145.1 Intron XP_011524447.1
XM_011526147.2 Intron XP_011524449.1
XM_011526148.2 Intron XP_011524450.1
XM_011526149.2 Intron XP_011524451.1
XM_011526150.2 Intron XP_011524452.1
XM_011526151.2 Intron XP_011524453.1
XM_011526152.2 Intron XP_011524454.1

View Full Product Details