Product Details

SNP ID
rs73557743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40316720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGAGGCTAGTTCAGATCTGCAA[C/T]GGACTCATCACCAGTATTTGCACTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf47 PubMed Links

Gene Details

Gene
C19orf47
Gene Name
chromosome 19 open reading frame 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256440.1 5767 Intron NP_001243369.1
NM_001256441.1 5767 Intron NP_001243370.1
XM_005258520.2 5767 Intron XP_005258577.1
XM_011526460.2 5767 Intron XP_011524762.1
XM_017026291.1 5767 Intron XP_016881780.1
XM_017026292.1 5767 UTR 3 XP_016881781.1
XM_017026293.1 5767 UTR 3 XP_016881782.1
XM_017026294.1 5767 Intron XP_016881783.1
XM_017026295.1 5767 Intron XP_016881784.1

View Full Product Details