Product Details
- SNP ID
-
rs73573953
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:48497387 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCCTGACTGTGCCCCGCAGCCT[C/T]CTCACCTTTGATCTGCTCGCTGTTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LMTK3
PubMed Links
Gene Details
- Gene
- LMTK3
- Gene Name
- lemur tyrosine kinase 3
View Full Product Details