Product Details
- SNP ID
-
rs73070545
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:50338102 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAAGCATCAAATCTTAATTTCACT[C/T]TTTTTTCCTGCAACCTCTGCCTCCT
- Phenotype
-
MIM: 176264
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KCNC3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111748895] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KCNC3
- Gene Name
- potassium voltage-gated channel subfamily C member 3
There are no transcripts associated with this gene.
- Gene
- NAPSB
- Gene Name
- napsin B aspartic peptidase, pseudogene
There are no transcripts associated with this gene.
View Full Product Details