Product Details

SNP ID
rs73054871
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51694172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGAGAGGGAATAGAGACCCAGA[A/G]AGGGGAGGATGGAGACTCAGAGGGT
Phenotype
MIM: 611191 MIM: 614510 MIM: 611250
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR125A PubMed Links

Gene Details

Gene
MIR125A
Gene Name
microRNA 125a
There are no transcripts associated with this gene.

Gene
MIR99B
Gene Name
microRNA 99b
There are no transcripts associated with this gene.

Gene
MIRLET7E
Gene Name
microRNA let-7e
There are no transcripts associated with this gene.

Gene
SPACA6
Gene Name
sperm acrosome associated 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316972.1 Intron NP_001303901.1
NM_001316994.1 Intron NP_001303923.1
XM_017026299.1 Intron XP_016881788.1
XM_017026300.1 Intron XP_016881789.1
XM_017026301.1 Intron XP_016881790.1
Gene
SPACA6P-AS
Gene Name
SPACA6P antisense RNA
There are no transcripts associated with this gene.

View Full Product Details