Product Details

SNP ID
rs73065212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57433318 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGTTATGTGGGATGTTGTCTGG[C/T]TTCCTGGCCTTGTTGCTAGACATAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF749 PubMed Links

Gene Details

Gene
ZNF749
Gene Name
zinc finger protein 749
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001023561.3 Intron NP_001018855.2
NM_001321952.1 Intron NP_001308881.1
NM_001321953.1 Intron NP_001308882.1
NM_001321954.1 Intron NP_001308883.1
XM_011526956.2 Intron XP_011525258.1
XM_017026802.1 Intron XP_016882291.1

View Full Product Details