Product Details

SNP ID
rs73066544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57701877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAACTTCCCACATTCACTGCACT[C/T]ATAAGGCCTCTCCCCAGTGTGAACT
Phenotype
MIM: 604085
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF154 PubMed Links

Gene Details

Gene
ZNF154
Gene Name
zinc finger protein 154
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085384.2 1312 Missense Mutation AAG,GAG K358E NP_001078853.1

View Full Product Details