Product Details
- SNP ID
-
rs73928330
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:35851795 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCACTCACCTTCAGTCAGCAGCC[C/G]CAGGAGCAGGAGAGAAGCCCTGAGC
- Phenotype
-
MIM: 607762
MIM: 602716
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KIRREL2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73928331] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KIRREL2
- Gene Name
- kin of IRRE like 2 (Drosophila)
- Gene
- NPHS1
- Gene Name
- NPHS1, nephrin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004646.3 |
199 |
Missense Mutation |
CGG,GGG |
R15G |
NP_004637.1 |
View Full Product Details