Product Details
- SNP ID
-
rs73624816
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:37186940 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATTCTGTGATGCCTGAAAAGAGA[C/T]GATACTTGAAAAAAGGATTTTCCAC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF585B
PubMed Links
Gene Details
- Gene
- ZNF585B
- Gene Name
- zinc finger protein 585B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152279.3 |
849 |
Silent Mutation |
TCA,TCG |
S199S |
NP_689492.3 |
View Full Product Details