Product Details

SNP ID
rs73520564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2333843 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGGCTGTTCCTTCTCCCTCCTG[C/T]GAAGTGGGAGCTGCTGTGCTGGACT
Phenotype
MIM: 607287 MIM: 608239
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LSM7 PubMed Links

Gene Details

Gene
LSM7
Gene Name
LSM7 homolog, U6 small nuclear RNA and mRNA degradation associated
There are no transcripts associated with this gene.

Gene
SPPL2B
Gene Name
signal peptide peptidase like 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077238.1 Intron NP_001070706.1
NM_152988.2 Intron NP_694533.1
XM_011528138.2 Intron XP_011526440.1
XM_011528139.1 Intron XP_011526441.1
XM_011528140.1 Intron XP_011526442.1
XM_017027016.1 Intron XP_016882505.1
XM_017027017.1 Intron XP_016882506.1
XM_017027018.1 Intron XP_016882507.1
XM_017027019.1 Intron XP_016882508.1

View Full Product Details