Product Details
- SNP ID
-
rs73523282
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:3889681 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCATAAGCTTTGGAATCGGACGCCC[A/G]TGGTTCAAATCCCAATTCCCCAGCA
- Phenotype
-
MIM: 608179
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATCAY
PubMed Links
Gene Details
- Gene
- ATCAY
- Gene Name
- ATCAY, caytaxin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_033064.4 |
|
Intron |
|
|
NP_149053.1 |
View Full Product Details