Product Details

SNP ID
rs73015182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10257176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCGTGGAACCTGAACTCCGGACC[G/T]CGGCCTTTGTGTGGAACCTGAACTC
Phenotype
MIM: 611823
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPL4 PubMed Links

Gene Details

Gene
MRPL4
Gene Name
mitochondrial ribosomal protein L4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015956.2 Intron NP_057040.2
NM_146387.1 Intron NP_666499.1
NM_146388.1 Intron NP_666500.1
XM_011528045.2 Intron XP_011526347.1

View Full Product Details