Product Details

SNP ID
rs71338625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17315963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGACAGAAAAGACAAACATCCTC[C/T]TGCGCTACCTGCATCAGCAATGGGA
Phenotype
MIM: 610216 MIM: 611840
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANO8 PubMed Links

Gene Details

Gene
ANO8
Gene Name
anoctamin 8
There are no transcripts associated with this gene.

Gene
DDA1
Gene Name
DET1 and DDB1 associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024050.5 293 Silent Mutation CTG,TTG L56L NP_076955.1
Gene
MRPL34
Gene Name
mitochondrial ribosomal protein L34
There are no transcripts associated with this gene.

View Full Product Details