Product Details

SNP ID
rs73006398
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9612771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCACTGGCTCTAATTGTTCAAA[A/G]AAAGAAAACTGTAGGGTTTTTTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF561 PubMed Links

Gene Details

Gene
ZNF561
Gene Name
zinc finger protein 561
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152289.2 Intron NP_689502.2
XM_005260150.2 Intron XP_005260207.1
XM_005260151.2 Intron XP_005260208.1
XM_006722938.2 Intron XP_006723001.1
XM_011528433.2 Intron XP_011526735.1
XM_011528434.2 Intron XP_011526736.1
XM_011528436.2 Intron XP_011526738.1
XM_011528437.2 Intron XP_011526739.1
XM_017027478.1 Intron XP_016882967.1
XM_017027479.1 Intron XP_016882968.1
XM_017027480.1 Intron XP_016882969.1
XM_017027481.1 Intron XP_016882970.1
XM_017027482.1 Intron XP_016882971.1
Gene
ZNF561-AS1
Gene Name
ZNF561 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details