Product Details

SNP ID
rs73296033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46368467 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAAGTTCTCTTGGGTAGCTGACCA[C/T]AGAGAAACTGAAAAGGGGAGAATTT
Phenotype
MIM: 606421
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ELMO2 PubMed Links

Gene Details

Gene
ELMO2
Gene Name
engulfment and cell motility 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318253.1 Intron NP_001305182.1
NM_133171.4 Intron NP_573403.1
NM_182764.2 Intron NP_877496.1
XM_005260496.3 Intron XP_005260553.1
XM_005260498.3 Intron XP_005260555.1
XM_005260499.3 Intron XP_005260556.1
XM_005260500.3 Intron XP_005260557.1
XM_005260501.4 Intron XP_005260558.1
XM_006723854.3 Intron XP_006723917.1
XM_017028009.1 Intron XP_016883498.1
XM_017028010.1 Intron XP_016883499.1
XM_017028011.1 Intron XP_016883500.1
Gene
SLC35C2
Gene Name
solute carrier family 35 member C2
There are no transcripts associated with this gene.

View Full Product Details