Product Details

SNP ID
rs72661177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:47509025 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAATGCTCTTTTACTTAAATTTT[C/G]TTGATTGAACATAAATGAATTAAGC
Phenotype
MIM: 601937
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NCOA3 PubMed Links

Gene Details

Gene
NCOA3
Gene Name
nuclear receptor coactivator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174087.1 Intron NP_001167558.1
NM_001174088.1 Intron NP_001167559.1
NM_006534.3 Intron NP_006525.2
NM_181659.2 Intron NP_858045.1

View Full Product Details