Product Details
- SNP ID
-
rs73598395
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:50082842 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTTTAGACATCATTAGGCGCCGAA[A/G]CTCTTGCAGGACAACTTTGATGCTA
- Phenotype
-
MIM: 602995
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM189-UBE2V1
PubMed Links
Gene Details
- Gene
- TMEM189-UBE2V1
- Gene Name
- TMEM189-UBE2V1 readthrough
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_199203.2 |
746 |
Missense Mutation |
CTT,TTT |
L347F |
NP_954673.1 |
- Gene
- UBE2V1
- Gene Name
- ubiquitin conjugating enzyme E2 V1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001032288.2 |
746 |
Missense Mutation |
CTT,TTT |
L124F |
NP_001027459.1 |
NM_001257393.1 |
746 |
Missense Mutation |
CTT,TTT |
L147F |
NP_001244322.1 |
NM_001257394.1 |
746 |
Missense Mutation |
CTT,TTT |
L80F |
NP_001244323.1 |
NM_001257395.1 |
746 |
Missense Mutation |
CTT,TTT |
L121F |
NP_001244324.1 |
NM_001257396.1 |
746 |
Missense Mutation |
CTT,TTT |
L82F |
NP_001244325.1 |
NM_001257397.1 |
746 |
Missense Mutation |
CTT,TTT |
L82F |
NP_001244326.1 |
NM_001257398.1 |
746 |
Missense Mutation |
CTT,TTT |
L82F |
NP_001244327.1 |
NM_001257399.1 |
746 |
Missense Mutation |
CTT,TTT |
L82F |
NP_001244328.1 |
NM_001282575.2 |
746 |
Missense Mutation |
CTT,TTT |
L68F |
NP_001269504.1 |
NM_001282576.1 |
746 |
Missense Mutation |
CTT,TTT |
L82F |
NP_001269505.1 |
NM_001282577.2 |
746 |
Missense Mutation |
CTT,TTT |
L68F |
NP_001269506.1 |
NM_001282578.2 |
746 |
Missense Mutation |
CTT,TTT |
L68F |
NP_001269507.1 |
NM_001282579.1 |
746 |
Missense Mutation |
CTT,TTT |
L82F |
NP_001269508.1 |
NM_001282580.1 |
746 |
Intron |
|
|
NP_001269509.1 |
NM_021988.5 |
746 |
Missense Mutation |
CTT,TTT |
L147F |
NP_068823.2 |
NM_022442.5 |
746 |
Missense Mutation |
CTT,TTT |
L80F |
NP_071887.1 |
NM_199144.2 |
746 |
Missense Mutation |
CTT,TTT |
L147F |
NP_954595.1 |
View Full Product Details