Product Details

SNP ID
rs73908164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:45611090 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAATGGCCAGACGACATTTCAT[A/G]TTTCCAATGAACCCTTGAAATCTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WFDC9 PubMed Links

Gene Details

Gene
WFDC9
Gene Name
WAP four-disulfide core domain 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147198.3 Intron NP_671731.1

View Full Product Details