Product Details
- SNP ID
-
rs73069132
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:5756397 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAAATATTAAGTCTTTTGATCCA[C/T]GAACATGGGATAGCTTTCCATTCCT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C20orf196
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs372628016] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C20orf196
- Gene Name
- chromosome 20 open reading frame 196
View Full Product Details