Product Details

SNP ID
rs73343142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:25395356 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTAACCATTATTAGGGAAATACA[C/T]CAAAATGTTAACATTGGTTGTATTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00158 PubMed Links

Gene Details

Gene
LINC00158
Gene Name
long intergenic non-protein coding RNA 158
There are no transcripts associated with this gene.

View Full Product Details