Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001003696.1 | 417 | Intron | NP_001003696.1 | ||
NM_001003697.1 | 417 | Intron | NP_001003697.1 | ||
NM_001003701.1 | 417 | UTR 5 | NP_001003701.1 | ||
NM_001003703.1 | 417 | UTR 5 | NP_001003703.1 | ||
NM_001320266.1 | 417 | UTR 5 | NP_001307195.1 | ||
NM_001320267.1 | 417 | UTR 5 | NP_001307196.1 | ||
NM_001685.4 | 417 | UTR 5 | NP_001676.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001197297.1 | 417 | Intron | NP_001184226.1 | ||
NM_002040.3 | 417 | UTR 5 | NP_002031.2 | ||
XM_005260938.4 | 417 | Intron | XP_005260995.1 | ||
XM_011529520.2 | 417 | UTR 5 | XP_011527822.1 | ||
XM_011529521.2 | 417 | UTR 5 | XP_011527823.1 | ||
XM_017028313.1 | 417 | Intron | XP_016883802.1 |