Product Details
- SNP ID
-
rs73169353
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:39577981 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGGGATCCAATGAGAATTCTGGG[G/T]CAAGCATGGACAAACTAAGTGCTGA
- Phenotype
-
MIM: 608230
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CACNA1I
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs71799331] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CACNA1I
- Gene Name
- calcium voltage-gated channel subunit alpha1 I
View Full Product Details