Product Details

SNP ID
rs73443974
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:49920668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGCACTGGGTGTCTATCAGGCTG[A/G]GCACATGGCCTTGAGCCCCAATGGG
Phenotype
MIM: 607144 MIM: 607171
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALG12 PubMed Links

Gene Details

Gene
ALG12
Gene Name
ALG12, alpha-1,6-mannosyltransferase
There are no transcripts associated with this gene.

Gene
CRELD2
Gene Name
cysteine rich with EGF like domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135101.2 Intron NP_001128573.1
NM_001284317.1 Intron NP_001271246.1
NM_001284318.1 Intron NP_001271247.1
NM_024324.4 Intron NP_077300.3
XM_005261737.2 Intron XP_005261794.1
XM_005261738.4 Intron XP_005261795.1
XM_011530372.2 Intron XP_011528674.1
XM_017028938.1 Intron XP_016884427.1

View Full Product Details