Product Details

SNP ID
rs73490430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55038856 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGAATAAGGAGGTAGCACTGAC[A/G]TCAAATAAAATATAGAGACAAATCT
Phenotype
MIM: 301300
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALAS2 PubMed Links

Gene Details

Gene
ALAS2
Gene Name
5'-aminolevulinate synthase 2
There are no transcripts associated with this gene.

Gene
PAGE2B
Gene Name
PAGE family member 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015038.2 Intron NP_001015038.1
XM_011530785.2 Intron XP_011529087.1
XM_011530786.2 Intron XP_011529088.1
XM_011530787.2 Intron XP_011529089.1
XM_017029513.1 Intron XP_016885002.1

View Full Product Details