Product Details

SNP ID
rs72616065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:18442421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATGGTCGTCTCCTCTCTCCCTT[C/T]AGGTACTCTCCTTAGTTTTTGAGCC
Phenotype
MIM: 300203
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDKL5 PubMed Links

Gene Details

Gene
CDKL5
Gene Name
cyclin dependent kinase like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037343.1 85 UTR 5 NP_001032420.1
NM_001323289.1 85 Intron NP_001310218.1
NM_003159.2 85 Intron NP_003150.1

View Full Product Details