Product Details

SNP ID
rs73201438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24167653 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTTTTAAAGAAAAATTCAGTAG[A/G]TGGGGTGGAGATCAAATTAGCTAGC
Phenotype
MIM: 314980
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFX PubMed Links

Gene Details

Gene
ZFX
Gene Name
zinc finger protein, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178084.1 Intron NP_001171555.1
NM_001178085.1 Intron NP_001171556.1
NM_001178086.1 Intron NP_001171557.1
NM_001178095.1 Intron NP_001171566.1
NM_003410.3 Intron NP_003401.2
XM_005274591.4 Intron XP_005274648.1
XM_005274592.3 Intron XP_005274649.1
XM_005274594.4 Intron XP_005274651.1
XM_006724513.3 Intron XP_006724576.1
XM_006724515.3 Intron XP_006724578.1
XM_011545578.1 Intron XP_011543880.1
XM_011545579.1 Intron XP_011543881.1
XM_011545581.2 Intron XP_011543883.1
XM_011545583.2 Intron XP_011543885.1
XM_017029788.1 Intron XP_016885277.1
XM_017029789.1 Intron XP_016885278.1
XM_017029790.1 Intron XP_016885279.1
XM_017029791.1 Intron XP_016885280.1
XM_017029792.1 Intron XP_016885281.1
XM_017029793.1 Intron XP_016885282.1
XM_017029794.1 Intron XP_016885283.1
XM_017029795.1 Intron XP_016885284.1
XM_017029796.1 Intron XP_016885285.1
XM_017029797.1 Intron XP_016885286.1
XM_017029798.1 Intron XP_016885287.1
XM_017029799.1 Intron XP_016885288.1
XM_017029800.1 Intron XP_016885289.1
XM_017029801.1 Intron XP_016885290.1

View Full Product Details