Product Details

SNP ID
rs73239352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:133024685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACAAAGGCAAGGCATACATTAA[C/T]ATGAAAAACACAAAAATGTAATCAA
Phenotype
MIM: 300309
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP26 PubMed Links

Gene Details

Gene
USP26
Gene Name
ubiquitin specific peptidase 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031907.1 3674 Intron NP_114113.1
XM_017029892.1 3674 UTR 3 XP_016885381.1

View Full Product Details