Product Details

SNP ID
rs72625375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.Y:19708042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACCGGCTACGCTTGGTGCTGTC[T/C]GAGCCAGCGTCTGCACACGGGCAAA
Phenotype
MIM: 426000
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
KDM5D PubMed Links

Gene Details

Gene
KDM5D
Gene Name
lysine demethylase 5D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146705.1 3756 Silent Mutation TCA,TCG S1128S NP_001140177.1
NM_001146706.1 3756 Silent Mutation TCA,TCG S1040S NP_001140178.1
NM_004653.4 3756 Silent Mutation TCA,TCG S1097S NP_004644.2
XM_005262560.2 3756 Silent Mutation TCA,TCG S1052S XP_005262617.1
XM_005262561.2 3756 Silent Mutation TCA,TCG S1020S XP_005262618.1
XM_011531468.2 3756 Silent Mutation TCA,TCG S1071S XP_011529770.1

View Full Product Details