Product Details

SNP ID
rs72779355
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:19911243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGAAAGTGCAACCAACAAATCA[C/G]AGTAAGGGGCTAGAGGCATGACAAT
Phenotype
MIM: 613602
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TTC32 PubMed Links

Gene Details

Gene
TTC32
Gene Name
tetratricopeptide repeat domain 32
There are no transcripts associated with this gene.

Gene
WDR35
Gene Name
WD repeat domain 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006657.1 4661 UTR 3 NP_001006658.1
NM_020779.3 4661 UTR 3 NP_065830.2
XM_011533007.2 4661 UTR 3 XP_011531309.1

View Full Product Details