Product Details

SNP ID
rs72970768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:85480698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTAGTGCTTTTATAATCCCTGA[G/T]AGATAAAATGACCTTTCTTCCAAAT
Phenotype
MIM: 610586
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP24 PubMed Links

Gene Details

Gene
ARHGAP24
Gene Name
Rho GTPase activating protein 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025616.2 Intron NP_001020787.2
NM_001042669.1 Intron NP_001036134.1
NM_001287805.1 Intron NP_001274734.1
NM_031305.2 Intron NP_112595.2
XM_011532300.2 Intron XP_011530602.1
XM_017008679.1 Intron XP_016864168.1

View Full Product Details