Product Details

SNP ID
rs73829239
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:80185673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGCTGTCCCCTTGCGAGGGCAC[G/T]AGGACAACAGGAGGGGCGCACCTGC
Phenotype
MIM: 616639
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PRDM8 PubMed Links

Gene Details

Gene
PRDM8
Gene Name
PR domain 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099403.1 Intron NP_001092873.1
NM_020226.3 Intron NP_064611.3
XM_005263144.2 Intron XP_005263201.1
XM_005263145.2 Intron XP_005263202.1
XM_005263146.4 Intron XP_005263203.1
XM_011532133.2 Intron XP_011530435.1
XM_011532135.2 Intron XP_011530437.1
XM_011532140.2 Intron XP_011530442.1
XM_011532141.2 Intron XP_011530443.1
XM_017008468.1 Intron XP_016863957.1
XM_017008469.1 Intron XP_016863958.1
XM_017008470.1 Intron XP_016863959.1

View Full Product Details